2-Chloro-4-fluorophenylacetonitrile ≥95%
Supplier: Matrix Scientific
075088-100G
075088-25G
102539-706EA
553.85
USD
102539-706
102833-774
2-Chloro-4-fluorophenylacetonitrile ≥95%
2-Chloro-4-fluorophenylacetonitrile
Hazard: Toxic
Formula:
C₈H₅ClFN Melting Pt: 67…69 °C |
MDL Number:
MFCD00236029 CAS Number: 75279-56-0 UN: 3439 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...