tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 4-aminopiperidine-1-carboxylate hydrochloride, 4-Amino-1-Boc-piperidine hydrochloride, tert-Butyl 4-amino-1-piperidinecarboxylate hydrochloride
075087-1G
075087-5G
075087-10G
102539-698EA
178.46
USD
102539-698
102539-700
102539-702
tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride ≥95%
tert-Butyl-4-aminopiperidine-1-carboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₁ClN₂O₂ MW: 236.7400 g/mol |
MDL Number:
MFCD01321014 CAS Number: 189819-75-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...