Diethyl-2-(aminomethylene)malonate ≥95%

Supplier: Matrix Scientific

Synonyms: Diethyl 2-(aminomethylene)malonate

075082-5G 075082-25G
102539-672EA 93.85 USD
102539-672 102664-034
Diethyl-2-(aminomethylene)malonate ≥95%
Diethyl-2-(aminomethylene)malonate

Hazard: Irritant

Formula: C₈H₁₃NO₄
MW: 187.20 g/mol
MDL Number: MFCD01075695
CAS Number: 6296-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR