3-Acetyl-2,5-dimethylthiophene ≥95%
Supplier: Matrix Scientific
075079-5G
075079-10G
075079-100G
102539-654EA
178.46
USD
102539-654
102539-656
102539-658
3-Acetyl-2,5-dimethylthiophene ≥95%
3-Acetyl-2,5-dimethylthiophene
Hazard: Irritant
Formula:
C₈H₁₀OS MW: 154.23 g/mol Boiling Pt: 126°C/28 Torr |
MDL Number:
MFCD00009763 CAS Number: 2530-10-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...