3,4-Diaminobenzoic acid ≥95%
Supplier: Matrix Scientific
075078-10G
075078-100G
102539-650EA
29.23
USD
102539-650
102539-652
3,4-Diaminobenzoic acid ≥95%
3,4-Diaminobenzoic acid
Hazard: Irritant
Formula:
C₇H₈N₂O₂ Melting Pt: ∼210 °C (dec.) |
MDL Number:
MFCD00007726 CAS Number: 619-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...