(S)-3-Amino-3-(2-methylphenyl)propionic acid ≥95%

Supplier: Matrix Scientific

075077-1G 075077-5G
102539-646EA 283.08 USD
102539-646 102539-648
(S)-3-Amino-3-(2-methylphenyl)propionic acid ≥95%
(S)-3-Amino-3-(2-methylphenyl)propionic acid

Hazard: Irritant

Formula: C₁₀H₁₃NO₂
MW: 179.2200 g/mol
MDL Number: MFCD04113679
CAS Number: 736131-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR