2-Amino-5-hydroxybenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxyanthranilic acid

075076-5G 075076-10G 075076-100G
102539-640EA 55.38 USD
102539-640 102539-642 102539-644
2-Amino-5-hydroxybenzoic acid ≥95%
2-Amino-5-hydroxybenzoic acid

Hazard: Irritant

Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00007870
CAS Number: 394-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR