3,4-Diaminobenzonitrile ≥95%

Supplier: Matrix Scientific

075073-5G 075073-25G
102539-626EA 69.23 USD
102539-626 102664-032
3,4-Diaminobenzonitrile ≥95%
3,4-Diaminobenzonitrile

Hazard: Irritant

Formula: C₇H₇N₃
Melting Pt: 144…148 °C
MDL Number: MFCD00723901
CAS Number: 17626-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR