(R)-Dimethylpyrrolidin-3-ylamine dihydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-Dimethylpyrrolidin-3-yl-amine dihydrochloride
075072-1G
075072-5G
075072-10G
102539-618EA
533.85
USD
102539-618
102539-620
102539-622
(R)-Dimethylpyrrolidin-3-ylamine dihydrochloride ≥95%
(R)-Dimethylpyrrolidin-3-ylamine dihydrochloride
Hazard: Irritant
Formula:
C₆H₁₆Cl₂N₂ MW: 187.11 g/mol |
MDL Number:
MFCD08704347 CAS Number: 864448-61-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...