Quinaldine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methylquinoline

075071-10G 075071-100G
102539-614EA 72.31 USD
102539-614 102539-616
Quinaldine ≥95%
Quinaldine

Hazard: Irritant

Formula: C₁₀H₉N
MW: 143.19 g/mol
MDL Number: MFCD00006756
CAS Number: 91-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR