(R)-N-Boc-piperidine-2-methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
(R)-N-Boc-Piperidine-2-methanol, tert-Butyl (R)-2-(hydroxymethyl)piperidine-1-carboxylate
075070-1G
075070-5G
102539-610EA
533.85
USD
102539-610
102539-612
(R)-N-Boc-piperidine-2-methanol ≥95%
(R)-N-Boc-piperidine-2-methanol
Hazard: Irritant
Formula:
C₁₁H₂₁NO₃ MW: 215.2900 g/mol |
MDL Number:
MFCD04973120 CAS Number: 134441-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...