Boc-D-Abu-OH DCHA ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-2-Boc-aminobutyric acid dicyclohexylammonium salt, Boc-D-Abu-OH.DCHA, Boc-D-Abu-OH·DCHA

075069-1G 075069-5G 075069-10G
102539-604EA 213.85 USD
102539-604 102539-606 102539-608
Boc-D-Abu-OH DCHA ≥95%
Boc-D-Abu-OH DCHA

Hazard: Irritant

Formula: C₂₁H₄₀N₂O₄
MW: 384.56 g/mol
MDL Number: MFCD00235886
CAS Number: 27494-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR