2-Hydroxybenzimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Benzimidazolinone, 2(1H)-Benzimidazolone, 2-Hydroxy-1H-benzimidazole

075067-100G 075067-25G
102539-598EA 292.31 USD
102539-598 102833-772
2-Hydroxybenzimidazole ≥95%
2-Hydroxybenzimidazole

Hazard: Irritant

Formula: C₇H₆N₂O
MW: 134.14 g/mol
Boiling Pt: 185…190 °C (0.01 Torr)
Melting Pt: 314…316 °C
MDL Number: MFCD00127894
CAS Number: 615-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR