5-Chloro-2-nitrotoluene ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-methylnitrobenzene

075066-5G 075066-10G 075066-100G
102539-588EA 46.15 USD
102539-588 102539-590 102539-592
5-Chloro-2-nitrotoluene ≥95%
5-Chloro-2-nitrotoluene

Hazard: Toxic

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 124…126 °C (16 mmHg)
Melting Pt: 23…25 °C
Density: 1.32
MDL Number: MFCD00024579
CAS Number: 5367-28-2
UN: 3457
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR