3,5-Dichlorobenzyl alcohol ≥95%

Supplier: Matrix Scientific

075065-5G 075065-10G 075065-100G
102539-582EA 98.46 USD
102539-582 102539-584 102539-586
3,5-Dichlorobenzyl alcohol ≥95%
3,5-Dichlorobenzyl alcohol

Hazard: Irritant

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
MDL Number: MFCD00004634
CAS Number: 60211-57-6
UN: 2250
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR