3,4,5-Trichlorophenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (3,4,5-Trichlorophenyl)boronic acid

075064-1G 075064-5G 075064-25G
102539-576EA 21.54 USD
102539-576 102539-578 102833-770
3,4,5-Trichlorophenylboronic acid ≥95%
3,4,5-Trichlorophenylboronic acid

Hazard: Irritant

Formula: C₆H₄BCl₃O₂
MW: 225.27 g/mol
MDL Number: MFCD03095126
CAS Number: 862248-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR