2-Amino-5-chlorobenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-chloro-1,3-benzothiazol-2-amine

075061-1G 075061-5G
102539-560EA 169.23 USD
102539-560 102539-562
2-Amino-5-chlorobenzothiazole ≥95%
2-Amino-5-chlorobenzothiazole

Hazard: Irritant

Formula: C₇H₅ClN₂S
MW: 184.65 g/mol
MDL Number: MFCD00205355
CAS Number: 20358-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR