4-Chloro-2-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

075057-10G 075057-100G
102539-538EA 72.31 USD
102539-538 102539-540
4-Chloro-2-nitrobenzoic acid ≥95%
4-Chloro-2-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄ClNO₄
MDL Number: MFCD00007214
CAS Number: 6280-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR