(R)-2,3-Dihydro-1H-inden-1-amine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (1R)-(-)-1-Aminoindane hydrochloride

075053-1G 075053-5G 075053-10G
102539-514EA 178.46 USD
102539-514 102539-516 102539-518
(R)-2,3-Dihydro-1H-inden-1-amine hydrochloride ≥95%
(R)-2,3-Dihydro-1H-inden-1-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClN
MW: 169.6513 g/mol
MDL Number: MFCD11100854
CAS Number: 10305-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR