Ethyl-4-methylpiperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

075052-250MG 075052-1G 075052-5G
102539-508EA 400 USD
102539-508 102539-510 102539-512
Ethyl-4-methylpiperidine-4-carboxylate ≥95%
Ethyl-4-methylpiperidine-4-carboxylate

Hazard: Irritant

Formula: C₉H₁₇NO₂
MDL Number: MFCD11045405
CAS Number: 103039-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR