5-(Cyanomethyl)-2-fluorobenzonitrile ≥95%

Supplier: Matrix Scientific

075051-1G 075051-5G
102539-504EA 710.77 USD
102539-504 102539-506
5-(Cyanomethyl)-2-fluorobenzonitrile ≥95%
5-(Cyanomethyl)-2-fluorobenzonitrile

Hazard: Irritant

Formula: C₉H₅FN₂
MW: 160.1500 g/mol
MDL Number: MFCD03094342
CAS Number: 519059-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR