4-Fluoro-3-nitrobenzotrifluoride ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Fluoro-3-Nitrobenzotrifluoride
075050-10G
075050-100G
102539-500EA
36.92
USD
102539-500
102539-502
4-Fluoro-3-nitrobenzotrifluoride ≥95%
4-Fluoro-3-nitrobenzotrifluoride
Hazard: Flammable
Formula:
C₇H₃F₄NO₂ MW: 209.1 g/mol Boiling Pt: 91-92°C/15mm Density: 1.494 Flash Pt: 33°C |
MDL Number:
MFCD00007059 CAS Number: 367-86-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...