Benzyltrimethylammonium tribromide ≥95%
Supplier: Matrix Scientific
Synonyms:
Mono(N,N,N-trimethyl-1-phenylmethanaminium) tribromide, Benzyl trimethyl ammonium tribromide
075049-5G
075049-10G
075049-100G
102539-494EA
23.08
USD
102539-494
102539-496
102539-498
Benzyltrimethylammonium tribromide ≥95%
Benzyltrimethylammonium tribromide
Hazard: Irritant/Hygroscopic
Formula:
C₁₀H₁₆Br₃N MW: 389.97 g/mol Melting Pt: 99…101 °C |
MDL Number:
MFCD00134423 CAS Number: 111865-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...