4-Chloro-3-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

075047-5G 075047-100G
102539-482EA 16.92 USD
102539-482 102539-486
4-Chloro-3-nitrobenzonitrile ≥95%
4-Chloro-3-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₃ClN₂O₂
MDL Number: MFCD00016987
CAS Number: 939-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR