(S)-2-Piperidinone-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(2S)-6-Oxopiperidine-2-carboxylic acid
075046-1G
075046-5G
102539-478EA
269.23
USD
102539-478
102539-480
(S)-2-Piperidinone-6-carboxylic acid ≥95%
(S)-2-Piperidinone-6-carboxylic acid
Hazard: Irritant
Formula:
C₆H₉NO₃ MW: 143.1406 g/mol |
MDL Number:
MFCD03839863 CAS Number: 34622-39-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...