4-Bromo-3,5-dimethylbenzonitrile ≥95%

Supplier: Matrix Scientific

075040-1G 075040-5G
102539-450EA 153.85 USD
102539-450 102539-452
4-Bromo-3,5-dimethylbenzonitrile ≥95%
4-Bromo-3,5-dimethylbenzonitrile

Hazard: Irritant

Formula: C₉H₈BrN
MW: 210.0700 g/mol
MDL Number: MFCD07369910
CAS Number: 75344-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR