Dimethyl itaconate ≥95%

Supplier: Matrix Scientific

Synonyms: Itaconic acid dimethyl ester, Methylenesuccinic acid dimethyl ester

075037-100G 075037-500G
102539-434EA 75.38 USD
102539-434 102539-436
Dimethyl itaconate ≥95%
Dimethyl itaconate

Hazard: Irritant

Formula: C₇H₁₀O₄
MDL Number: MFCD00008446
CAS Number: 617-52-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR