3-(Hydroxymethyl)indazole ≥95%

Supplier: Matrix Scientific

Synonyms: (1H-Indazol-3-yl)methanol

075035-1G 075035-5G
102539-424EA 200 USD
102539-424 102539-426
3-(Hydroxymethyl)indazole ≥95%
3-(Hydroxymethyl)indazole

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.17 g/mol
MDL Number: MFCD05260509
CAS Number: 64132-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR