O-Methylhydroxylamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride

075034-100G 075034-25G
102539-422EA 112.31 USD
102539-422 102551-938
O-Methylhydroxylamine hydrochloride ≥95%
O-Methylhydroxylamine hydrochloride

Hazard: Corrosive/Hygroscopic

Formula: CH₆ClNO
MW: 83.515 g/mol
Melting Pt: ∼ 148 °C (dec.)
MDL Number: MFCD00012951
CAS Number: 593-56-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR