2,6-Dinitroaniline ≥95%
Supplier: Matrix Scientific
075033-10G
075033-100G
102539-416EA
246.15
USD
102539-416
102539-418
2,6-Dinitroaniline ≥95%
2,6-Dinitroaniline
Hazard: Irritant
Formula:
C₆H₅N₃O₄ MW: 183.12 g/mol |
MDL Number:
MFCD00007148 CAS Number: 606-22-4 UN: 1596 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...