Boc-L-alanine benzyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Boc-Ala-OBzl, Boc-L-Alanine benzyl ester

075031-1G 075031-5G 075031-10G
102539-404EA 72.31 USD
102539-404 102539-406 102539-408
Boc-L-alanine benzyl ester ≥95%
Boc-L-alanine benzyl ester

Hazard: Irritant

Formula: C₁₅H₂₁NO₄
MW: 279.34 g/mol
MDL Number: MFCD00190775
CAS Number: 51814-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR