4-Carboxyphenylacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (4-Carboxyphenyl)acetic acid, 4-(Carboxymethyl)benzoic acid

075029-1G 075029-5G 075029-25G
102539-394EA 21.54 USD
102539-394 102539-396 102833-766
4-Carboxyphenylacetic acid ≥95%
4-Carboxyphenylacetic acid

Hazard: Irritant

Formula: C₉H₈O₄
MDL Number: MFCD01074834
CAS Number: 501-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR