4-Phenoxypyridine ≥95%

Supplier: Matrix Scientific

075028-5G 075028-10G 075028-100G
102539-388EA 284.62 USD
102539-388 102539-390 102539-392
4-Phenoxypyridine ≥95%
4-Phenoxypyridine

Hazard: Irritant

Formula: C₁₁H₉NO
MW: 171.2000 g/mol
MDL Number: MFCD00235157
CAS Number: 4783-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR