Cyclopropanecarbonyl chloride ≥95%

Supplier: Matrix Scientific

075027-100G 075027-500G
102539-386EA 184.62 USD
102539-386 102664-030
Cyclopropanecarbonyl chloride ≥95%
Cyclopropanecarbonyl chloride

Hazard: Flammable/Corrosive/Moisture Sensitive/Refrigerate

Formula: C₄H₅ClO
Boiling Pt: 119 °C
Density: 1.152
Flash Pt: 23 °C
MDL Number: MFCD00001277
CAS Number: 4023-34-1
UN: 2920
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR