(R)-3-Aminobutyric acid ≥95%

Supplier: Matrix Scientific

075025-1G 075025-5G
102539-376EA 76.92 USD
102539-376 102539-378
(R)-3-Aminobutyric acid ≥95%
(R)-3-Aminobutyric acid

Hazard: Irritant

Formula: C₄H₉NO₂
MW: 103.1198 g/mol
MDL Number: MFCD00211284
CAS Number: 3775-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR