Dibutylamine hydrochloride ≥95%

Supplier: Matrix Scientific

075024-10G 075024-100G
102539-372EA 27.69 USD
102539-372 102539-374
Dibutylamine hydrochloride ≥95%
Dibutylamine hydrochloride

Hazard: Irritant

Formula: C₈H₂₀ClN
MW: 165.71 g/mol
MDL Number: MFCD00034885
CAS Number: 6287-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR