1-Bromo-4-nitrobenzene ≥95%

Supplier: Matrix Scientific

075021-100G 075021-25G
102539-360EA 170.77 USD
102539-360 102664-028
1-Bromo-4-nitrobenzene ≥95%
1-Bromo-4-nitrobenzene

Hazard: Toxic

Formula: C₆H₄BrNO₂
Melting Pt: 124…127 °C
MDL Number: MFCD00007280
CAS Number: 586-78-7
UN: 3459
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR