1-Chloro-3,5-dibromobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene

075018-100G 075018-25G
102539-348EA 192.31 USD
102539-348 102833-764
1-Chloro-3,5-dibromobenzene ≥95%
1-Chloro-3,5-dibromobenzene

Hazard: Irritant

Formula: C₆H₃Br₂Cl
MW: 270.35
Melting Pt: 95…98 °C
MDL Number: MFCD00070765
CAS Number: 14862-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR