4-Acetylbenzonitrile ≥95%
Supplier: Matrix Scientific
075016-1G
075016-5G
075016-10G
102539-334EA
36.92
USD
102539-334
102539-336
102539-338
4-Acetylbenzonitrile ≥95%
4-Acetylbenzonitrile
Hazard: Irritant
Formula:
C₉H₇NO MW: 145.16 g/mol |
MDL Number:
MFCD00001825 CAS Number: 1443-80-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...