1,3-Difluoro-4-nitrobenzene ≥95%

Supplier: Avantor

Synonyms: 2,4-Difluoro-1-nitrobenzene, 2,4-Difluoronitrobenzene

075013-100G
102539-320EA 27.6 USD
102539-320 102539-322
1,3-Difluoro-4-nitrobenzene ≥95%
1,3-Difluoro-4-nitrobenzene

Hazard: Irritant

Formula: C₆H₃F₂NO₂
MW: 159.09 g/mol
MDL Number: MFCD00007050
CAS Number: 446-35-5
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR