N,N'-((1R,2R)-cyclohexane-1,2-diyl)dimethanesulfonamide ≥95%

Supplier: Matrix Scientific

075011-250MG 075011-1G
102539-310EA 446.15 USD
102539-310 102539-312
N,N'-((1R,2R)-cyclohexane-1,2-diyl)dimethanesulfonamide ≥95%
N,N'-((1R,2R)-cyclohexane-1,2-diyl)dimethanesulfonamide

Hazard: Irritant

Formula: C₈H₁₈N₂O₄S₂
MDL Number: MFCD03411210
CAS Number: 122833-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR