Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) ≥95%

Supplier: Matrix Scientific

Synonyms: N-Hydroxy sulphosuccinimide sodium salt, Hydroxy-2,5-dioxopyrrolidine-3-sulphonic acid sodium salt, N-Hydroxysulphosuccinimide sodium salt, Sulfo NHS, Sulpho-N-Hydroxysuccinimide

075007-1G 075007-5G 075007-25G
102539-292EA 69.23 USD
102539-292 102539-294 102664-026
Sulfo-NHS (N-Hydroxysulfosuccinimide sodium salt) ≥95%
Sulpho-NHS (N-Hydroxysulphosuccinimide sodium salt)

Hazard: Irritant/Moisture Sensitive/Refrigerate

Formula: C₄H₄NNaO₆S
MW: 217.13 g/mol
Melting Pt: 250 °C
MDL Number: MFCD00043100
CAS Number: 106627-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR