4-Chloroaniline ≥95%

Supplier: Matrix Scientific

075006-10G 075006-100G
102539-288EA 36.92 USD
102539-288 102539-290
4-Chloroaniline ≥95%
4-Chloroaniline

Hazard: Irritant

Formula: C₆H₆ClN
MW: 127.57 g/mol
MDL Number: MFCD00007835
CAS Number: 106-47-8
UN: 2018
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR