Boc-D-Pro-OH ≥95%

Supplier: Matrix Scientific

075004-5G 075004-100G
102539-278EA 15.38 USD
102539-278 102539-282
Boc-D-Pro-OH ≥95%
Boc-D-Pro-OH

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
Melting Pt: 133…135 °C
MDL Number: MFCD00063226
CAS Number: 37784-17-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR