2-Methylindole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-1H-indole

075003-10G 075003-100G
102539-274EA 36.92 USD
102539-274 102539-276
2-Methylindole ≥95%
2-Methylindole

Hazard: Irritant-Harmful/Refrigerate/Stench/Light Sensitive/Air Sensitive

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 272…273 °C
Melting Pt: 56…60 °C
Flash Pt: 141 °C
MDL Number: MFCD00005616
CAS Number: 95-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR