(±)-3-Amino-1,2-propanediol ≥95%

Supplier: Matrix Scientific

Synonyms: (±)-3-Amino-1,2-dihydroxy propane, 3-Amino-1,2-propanediol

075000-100G 075000-500G
102539-260EA 112.31 USD
102539-260 102833-760
(±)-3-Amino-1,2-propanediol ≥95%
(±)-3-Amino-1,2-propanediol

Hazard: Corrosive/Hygroscopic/Refrigerate

Formula: C₃H₉NO₂
MW: 91.11 g/mol
Boiling Pt: 264…266 °C (739 mmHg)
Density: 1.175
Flash Pt: 113 °C
MDL Number: MFCD00008140
CAS Number: 616-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR