4-(tert-Butyl)benzenesulfonamide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-tert-Butylbenzenesulphonamide

074999-25G 074999-100G
102833-756EA 110.77 USD
102833-756 102833-758
4-(tert-Butyl)benzenesulfonamide ≥95%
4-(tert-Butyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₅NO₂S
MW: 213.3000 g/mol
MDL Number: MFCD00068599
CAS Number: 6292-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR