3-Cyano-1H-indazole ≥95%

Supplier: Matrix Scientific

Synonyms: 1H-Indazole-3-carbonitrile

074996-1G 074996-5G
102539-238EA 126.15 USD
102539-238 102539-240
3-Cyano-1H-indazole ≥95%
3-Cyano-1H-indazole

Hazard: Irritant

Formula: C₈H₅N₃
MW: 143.15 g/mol
MDL Number: MFCD01860240
CAS Number: 50264-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR