3,3-Pentamethylene glutarimide ≥95%

Supplier: Matrix Scientific

Synonyms: Gabapentin Impurity 3

074994-1G 074994-5G 074994-10G
102539-226EA 143.08 USD
102539-226 102539-228 102539-230
3,3-Pentamethylene glutarimide ≥95%
3,3-Pentamethylene glutarimide

Hazard: Irritant

Formula: C₁₀H₁₅NO₂
MW: 181.235 g/mol
MDL Number: MFCD00023872
CAS Number: 1130-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR