Ethyl-1-methylpiperidine-4-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-methylpiperidine-4-carboxylate

074992-1G 074992-5G 074992-10G
102539-214EA 107.69 USD
102539-214 102539-216 102539-218
Ethyl-1-methylpiperidine-4-carboxylate ≥95%
Ethyl-1-methylpiperidine-4-carboxylate

Hazard: Irritant

Formula: C₉H₁₇NO₂
MW: 171.2400 g/mol
MDL Number: MFCD00130015
CAS Number: 24252-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR